Learn More
Grainger Deluxe Rotary Cutter, 7 1/4 in, Tungsten, 2 1/4 in Blade Diameter
Catalog No. NC1772175
Encompass_Preferred
Squeezing the handle of these rotary cutters extends the blade for cutting, and releasing the handle retracts the blade so that its edges are not exposed. This helps prevent injuries when the cutter is not in use.
Catalog No. NC1772175 Supplier Grainger Supplier No. 2CJU9
Squeezing the handle of these rotary cutters extends the blade for cutting, and releasing the handle retracts the blade so that its edges are not exposed. This helps prevent injuries when the cutter is not in use.