Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7,10-dimethyl-2,4-dioxobenzo[g]pteridine-8-carbaldehyde | 37854-59-4 | MFCD09038552 | 98.0% | 270.24 g/mol | C13H10N4O3 | 5 MG
SDP

Catalog No. 5000264773
Encompass_Preferred

Ro 08-2750 is a small-molecule research reagent that functions as a non-peptidic, reversible nerve growth factor (NGF) inhibitor and an inhibitor of Musashi (MSI) RNA-protein interactions. It is supplied as a high-purity solid for use in cellular and biochemical assays and is typically prepared in DMSO for in vitro studies.

  • Non-peptide, reversible NGF inhibitor with ~1 μM potency.
  • Also inhibits MSI RNA-binding activity (IC50 ≈ 2.7 μM).
  • High chemical purity (≈98%).
  • Molecular weight 270.24 g/mol; formula C13H10N4O3.
  • Supplied as a powder suitable for solution preparation.
  • Soluble in DMSO at approximately 4 mg/mL; may require warming and sonication.
  • Recommended storage: powder at -20°C for long-term stability.

Catalog No. 50-002-64773 Supplier Medchemexpress LLC Supplier No. HY1084665MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ro 08-2750 is a small-molecule research reagent that functions as a non-peptidic, reversible nerve growth factor (NGF) inhibitor and an inhibitor of Musashi (MSI) RNA-protein interactions. It is supplied as a high-purity solid for use in cellular and biochemical assays and is typically prepared in DMSO for in vitro studies.

  • Non-peptide, reversible NGF inhibitor with ~1 μM potency.
  • Also inhibits MSI RNA-binding activity (IC50 ≈ 2.7 μM).
  • High chemical purity (≈98%).
  • Molecular weight 270.24 g/mol; formula C13H10N4O3.
  • Supplied as a powder suitable for solution preparation.
  • Soluble in DMSO at approximately 4 mg/mL; may require warming and sonication.
  • Recommended storage: powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.