Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7,12-DMBA; 7,12-Dimethylbenzanthracene | 57-97-6 | 99.9% | 256.35 | 250 MG
SDP

Catalog No. 5000453335
Encompass_Preferred

DMBA (7,12-Dimethylbenzanthracene) is a polycyclic aromatic hydrocarbon (PAH) known for its carcinogenic activity. It is widely utilized in research to induce tumor formation in various rodent models, serving as a key tool in cancer studies.

  • Carcinogenic polycyclic aromatic hydrocarbon
  • Induces tumor formation in rodent models
  • Targets c-Myc
  • Inhibits apoptosis pathway
  • Solid, light yellow to yellow appearance
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • Solvent storage: -80°C for 6 months; -20°C for 1 month
  • Solubility in acetone, DMSO, ethanol, and corn oil

Catalog No. 50-004-53335 Supplier Medchemexpress LLC Supplier No. 250MGHYW011845
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DMBA (7,12-Dimethylbenzanthracene) is a polycyclic aromatic hydrocarbon (PAH) known for its carcinogenic activity. It is widely utilized in research to induce tumor formation in various rodent models, serving as a key tool in cancer studies.

  • Carcinogenic polycyclic aromatic hydrocarbon
  • Induces tumor formation in rodent models
  • Targets c-Myc
  • Inhibits apoptosis pathway
  • Solid, light yellow to yellow appearance
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • Solvent storage: -80°C for 6 months; -20°C for 1 month
  • Solubility in acetone, DMSO, ethanol, and corn oil

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.