Learn More
Ace Glass, Inc. 2ML SPECIFIC GRAVITY STOPPER-7/12
SDP
Catalog No. 50971881
Encompass_Preferred
Standard taper 7/12 stopper for Guy-Lussac type 2mL specific gravity bottle; unadjusted for calibration in the laboratory. When adjusted, this item is suitable for ASTM D 369.
Catalog No. 50-971-881 Supplier Ace Glass, Inc. Supplier No. 542005
Standard taper 7/12 stopper for Guy-Lussac type 2mL specific gravity bottle; unadjusted for calibration in the laboratory. When adjusted, this item is suitable for ASTM D 369.