Learn More
MSC Proto J1214BASD 7/16" 12 Point Offset Combination Wrench 15° Offset Angle, 7" Overall Length, Steel, Black Finish
Catalog No. 501299567
Encompass
Proto J1214BASD 7/16" 12 Point Offset Combination Wrench 15° Offset Angle, 7" Overall Length, Steel, Black Finish
Catalog No. 50-129-9567 Supplier MSC Supplier No. 64084429
Proto J1214BASD 7/16" 12 Point Offset Combination Wrench 15° Offset Angle, 7" Overall Length, Steel, Black Finish