Learn More
Ampco 7/16 in. Tip Insulated Non-Sparking Wrench
SDP
Catalog No. 19174130 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
5 -1/2 in.
Catalog No. Length (English)
19-174-130 5 -1/2 in.
1 options
Catalog No. 19-174-130 Supplier Ampco Supplier No. IWSB7/16
Add to Cart
Add to Cart
Insulated, non-magnetic, corrosion-resistant, and non-sparking wrench. Made in the USA.
  • Made in the USA.
  • HS Code: 8204.12.
  • Length: 5 and 1/2 in.
Specifications
Length (English) 5 -1/2 in.
Length (Metric) 140 mm
Material Aluminum Bronze
Product Type Insulated Wrench
WARNING: Cancer - www.P65Warnings.ca.gov