Learn More
Andwin Scientific 7/16" Laser Tough-Spots, 4,680/pk
SDP
Catalog No. NC1662623
Encompass_Preferred
Our patented Tough-Spots withstand both long term cryogenic storage conditions and conventional freezer storage. Cryogenic labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks.
Catalog No. NC1662623 Supplier Andwin Scientific Supplier No. SPOT5000
Our patented Tough-Spots withstand both long term cryogenic storage conditions and conventional freezer storage. Cryogenic labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks.