Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-{[2-(2,6-dioxopiperidin-3-yl)-1,3-dioxo-2,3-dihydro-1H-isoindol-4-yl]oxy}heptanoic acid | 2169266-69-5 | MFCD32063464 | 99.3% | C20H22N2O7 | 10MG
SDP

Catalog No. 5000261970
Encompass_Preferred

Thalidomide-O-C6-COOH is a thalidomide-based cereblon (CRBN) E3 ligase ligand-linker conjugate terminating in a six-carbon carboxylic acid. It is used as a building block in PROTAC synthesis and chemical biology research, providing a convenient carboxylic acid handle for linker attachment and efficient recruitment of CRBN.

  • Used in PROTAC design to recruit cereblon (CRBN) E3 ligase.
  • Thalidomide-based ligand with a six-carbon carboxylic acid linker.
  • High purity, 99.3% (reported).
  • Soluble ≥100 mg/mL in DMSO.
  • Off-white to light yellow solid suitable for synthesis.
  • Molecular formula C20H22N2O7; molecular weight 402.40 g·mol⁻1.
  • Storage recommendations provided for powder and solutions to maintain stability.

Catalog No. 50-002-61970 Supplier Medchemexpress LLC Supplier No. HY13095110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-O-C6-COOH is a thalidomide-based cereblon (CRBN) E3 ligase ligand-linker conjugate terminating in a six-carbon carboxylic acid. It is used as a building block in PROTAC synthesis and chemical biology research, providing a convenient carboxylic acid handle for linker attachment and efficient recruitment of CRBN.

  • Used in PROTAC design to recruit cereblon (CRBN) E3 ligase.
  • Thalidomide-based ligand with a six-carbon carboxylic acid linker.
  • High purity, 99.3% (reported).
  • Soluble ≥100 mg/mL in DMSO.
  • Off-white to light yellow solid suitable for synthesis.
  • Molecular formula C20H22N2O7; molecular weight 402.40 g·mol⁻1.
  • Storage recommendations provided for powder and solutions to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.