Learn More
Medchemexpress LLC 7-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)heptanoic acid | 2225940-50-9 | MFCD32063493 | 98.0% | C20H23N3O6 | 10MG
SDP
Catalog No. 5000260964
Encompass_Preferred
Pomalidomide-C6-COOH is a pomalidomide-derived cereblon (CRBN) ligand featuring a six-carbon carboxylic acid linker designed for use as a building block in PROTAC and targeted protein degradation research. It is supplied as a solid with high analytical purity suitable for medicinal chemistry and chemical biology applications.
  • Provides a carboxylic acid handle for linker attachment.
  • Designed for PROTAC design and targeted protein degradation studies.
  • High purity appropriate for research synthesis (≈98%).
  • Soluble in DMSO for assay and synthetic workups.
  • Supplied as a light yellow to yellow solid for ease of handling.
Catalog No. 50-002-60964 Supplier Medchemexpress LLC Supplier No. HY13188710MG
Add to Cart
Add to Cart
Pomalidomide-C6-COOH is a pomalidomide-derived cereblon (CRBN) ligand featuring a six-carbon carboxylic acid linker designed for use as a building block in PROTAC and targeted protein degradation research. It is supplied as a solid with high analytical purity suitable for medicinal chemistry and chemical biology applications.
  • Provides a carboxylic acid handle for linker attachment.
  • Designed for PROTAC design and targeted protein degradation studies.
  • High purity appropriate for research synthesis (≈98%).
  • Soluble in DMSO for assay and synthetic workups.
  • Supplied as a light yellow to yellow solid for ease of handling.