Learn More
Cayman Chemical 7 2 4DInItrophenoxy4meth 10mg
SDP
Catalog No. 502937751
Encompass_Preferred
A turn-on fluorescent probe for hydrogen sulfide; ex/em = 323/445 nm, respectively, in DMSO and fluorescence intensity increases in the presence of sulfur-containing molecules; selective for sulfur-containing molecules, including cysteine, GSH, homocysteine, and selenocysteine; not cytotoxic up to a concentration of 150 UG/ml
Catalog No. 50-293-7751 Supplier Cayman Chemical Supplier No. 2847210 MG
A turn-on fluorescent probe for hydrogen sulfide; ex/em = 323/445 nm, respectively, in DMSO and fluorescence intensity increases in the presence of sulfur-containing molecules; selective for sulfur-containing molecules, including cysteine, GSH, homocysteine, and selenocysteine; not cytotoxic up to a concentration of 150 UG/ml