Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-(2-hydroxypropan-2-yl)-4-[2-methyl-3-(4-oxo-3,4-dihydroquinazolin-3-yl)phenyl]-9H-carbazole-1-carb | 1231889-53-4 | MFCD30470964 | 99.1% | 502.56 | C31H26N4O3 | 2 MG
SDP

Catalog No. 5000245266
Encompass_Preferred

BMS-935177 is a small-molecule, reversible inhibitor of Bruton's tyrosine kinase (BTK) developed for research use. It is described in the medicinal chemistry literature as a potent and selective BTK inhibitor with an IC50 of approximately 3 nM and is supplied as a solid with high reported purity for biochemical and cellular studies.

  • Potent reversible inhibitor of Bruton's tyrosine kinase (IC50 ≈ 3 nM).
  • High selectivity for the BTK target in kinase panels.
  • Molecular formula C31H26N4O3 and molecular weight 502.56.
  • Supplied as a solid with reported purity around 99.1%.
  • Available in small pack sizes suitable for research (for example, 2 MG solid).
  • Soluble in DMSO and commonly provided as a 10 mM solution for assays.
  • Store powder at low temperature for long-term stability (e.g., -20°C).

Catalog No. 50-002-45266 Supplier Medchemexpress LLC Supplier No. HY1017932MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-935177 is a small-molecule, reversible inhibitor of Bruton's tyrosine kinase (BTK) developed for research use. It is described in the medicinal chemistry literature as a potent and selective BTK inhibitor with an IC50 of approximately 3 nM and is supplied as a solid with high reported purity for biochemical and cellular studies.

  • Potent reversible inhibitor of Bruton's tyrosine kinase (IC50 ≈ 3 nM).
  • High selectivity for the BTK target in kinase panels.
  • Molecular formula C31H26N4O3 and molecular weight 502.56.
  • Supplied as a solid with reported purity around 99.1%.
  • Available in small pack sizes suitable for research (for example, 2 MG solid).
  • Soluble in DMSO and commonly provided as a 10 mM solution for assays.
  • Store powder at low temperature for long-term stability (e.g., -20°C).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.