Learn More
Chemglass Life Sciences 7/25 Inner Joint, Full Length, on 6.35mm OD x 1.2mm W tubing
SDP
Supplier:  Chemglass Life Sciences CG10001
Encompass_Preferred
7/25 Inner Joint, Full Length, on 6.35mm OD x 1.2mm W tubing. Standard Taper ground Inner Joint, Full Length.
Catalog No. 50-153-0982