Learn More
Medchemexpress LLC 7-[3-(4-(2,3-dimethylphenyl)piperazin-1-yl)propoxy]-1H-quinolin-2-one hydrochloride | 1329509-60-5 | 99.0% | 427.97 g·mol⁻¹ | C24H30ClN3O2 | 1 MG
SDP
Catalog No. 5000241642
Encompass_Preferred
OPC 4392 hydrochloride is a small-molecule dopamine receptor modulator used in preclinical pharmacology research. It functions as a presynaptic dopamine autoreceptor agonist and a postsynaptic D2 receptor antagonist, and has been applied in animal studies to reverse reserpine-induced dopamine depletion and reduce apomorphine-induced behaviors.
  • Acts as presynaptic dopamine autoreceptor agonist and postsynaptic D2 antagonist.
  • Demonstrated efficacy in reversing reserpine-induced dopamine depletion in animal studies.
  • Inhibits apomorphine-induced stereotypic and climbing behaviors in rodent models.
  • High purity (98.99%) suitable for research applications.
  • White to off-white solid, convenient for handling and formulation.
  • Recommended sealed storage away from moisture; in solvent store at -80°C for long-term preservation.
Catalog No. 50-002-41642 Supplier Medchemexpress LLC Supplier No. HY106781A1MG
OPC 4392 hydrochloride is a small-molecule dopamine receptor modulator used in preclinical pharmacology research. It functions as a presynaptic dopamine autoreceptor agonist and a postsynaptic D2 receptor antagonist, and has been applied in animal studies to reverse reserpine-induced dopamine depletion and reduce apomorphine-induced behaviors.
  • Acts as presynaptic dopamine autoreceptor agonist and postsynaptic D2 antagonist.
  • Demonstrated efficacy in reversing reserpine-induced dopamine depletion in animal studies.
  • Inhibits apomorphine-induced stereotypic and climbing behaviors in rodent models.
  • High purity (98.99%) suitable for research applications.
  • White to off-white solid, convenient for handling and formulation.
  • Recommended sealed storage away from moisture; in solvent store at -80°C for long-term preservation.