Learn More
Apexbio Technology LLC 7,3',4'-Trihydroxyflavone 2150-11-0 5mg
SDP
Catalog No. 5000090144
Encompass_Preferred
73p4p-Trihydroxyflavone is a small-molecule inhibitor targeting cyclooxygenase-1 (COX-1) and pathways involved in osteoclastogenesis It is designed to inhibit oxidative radical formation and osteoclast differentiation thereby regulating inflammatory and bone resorption pathways 73p4p-Trihydroxyflavone exerts its biological activity primarily through free radical scavenging and downregulation of osteoclast marker genes In in vitro studies 73p4p-Trihydroxyflavone demonstrates inhibition of reactive oxygen species in rat kidney tissue homogenates and suppresses RANKL-induced osteoclastogenesis in mouse bone marrow-derived macrophages Based on these pharmacological properties 73p4p-Trihydroxyflavone holds research potential in biochemical pharmacological antimicrobial and bone metabolism studies
Catalog No. 50-000-90144 Supplier Apexbio Technology LLC Supplier No. N28455
73p4p-Trihydroxyflavone is a small-molecule inhibitor targeting cyclooxygenase-1 (COX-1) and pathways involved in osteoclastogenesis It is designed to inhibit oxidative radical formation and osteoclast differentiation thereby regulating inflammatory and bone resorption pathways 73p4p-Trihydroxyflavone exerts its biological activity primarily through free radical scavenging and downregulation of osteoclast marker genes In in vitro studies 73p4p-Trihydroxyflavone demonstrates inhibition of reactive oxygen species in rat kidney tissue homogenates and suppresses RANKL-induced osteoclastogenesis in mouse bone marrow-derived macrophages Based on these pharmacological properties 73p4p-Trihydroxyflavone holds research potential in biochemical pharmacological antimicrobial and bone metabolism studies