Learn More
Medchemexpress LLC 7-[3-(tert-butylamino)-2-hydroxy-propoxy]-3H-isobenzofuran-1-one hydrochloride | 55104-39-7 | 98.8% | 315.79 | C15H22ClNO4 | 10 MG
SDP
Catalog No. 5000242031
Encompass_Preferred
DL 071IT is a research-grade, non-selective beta-adrenergic receptor blocker with intrinsic sympathomimetic activity and weak membrane-stabilizing properties. It is supplied as a purified hydrochloride salt for in vitro and preclinical research use only.
  • High purity (98.8%).
  • Suitable for pharmacology and receptor profiling studies.
  • Exhibits intrinsic sympathomimetic activity.
  • Displays weak membrane-stabilizing properties.
  • Available in small research quantities for assay development.
  • Identifiable by CAS 55104-39-7 and formula C15H22ClNO4.
Catalog No. 50-002-42031 Supplier Medchemexpress LLC Supplier No. HY10666710MG
DL 071IT is a research-grade, non-selective beta-adrenergic receptor blocker with intrinsic sympathomimetic activity and weak membrane-stabilizing properties. It is supplied as a purified hydrochloride salt for in vitro and preclinical research use only.
  • High purity (98.8%).
  • Suitable for pharmacology and receptor profiling studies.
  • Exhibits intrinsic sympathomimetic activity.
  • Displays weak membrane-stabilizing properties.
  • Available in small research quantities for assay development.
  • Identifiable by CAS 55104-39-7 and formula C15H22ClNO4.