Learn More
Medchemexpress LLC 7-[(4,5-dihydro-2-methyl-4-oxo-2H-thieno[2,3-g]indol-7-yl)oxy]-N-(phenylsulfonyl)-9-heptanamide | 910562-18-4 | MFCD27572791 | 99.7% | 429.57 | 5 MG
SDP
Catalog No. 5000403265
Encompass_Preferred
Aganepag is a potent Prostanoid EP2 receptor agonist with an EC50 of 0.19 nM, showing no activity at the EP4 receptor. It is used in the research of wound healing, scar reduction, scar prevention, and wrinkle treatment and prevention.
  • Potent Prostanoid EP2 receptor agonist with an EC50 of 0.19 nM.
  • No activity at EP4 receptor.
  • Can be used in wound healing research.
  • Applicable for scar reduction and prevention studies.
  • Useful for wrinkle treatment and prevention research.
Catalog No. 50-004-03265 Supplier Medchemexpress LLC Supplier No. HY198645MG
Aganepag is a potent Prostanoid EP2 receptor agonist with an EC50 of 0.19 nM, showing no activity at the EP4 receptor. It is used in the research of wound healing, scar reduction, scar prevention, and wrinkle treatment and prevention.
  • Potent Prostanoid EP2 receptor agonist with an EC50 of 0.19 nM.
  • No activity at EP4 receptor.
  • Can be used in wound healing research.
  • Applicable for scar reduction and prevention studies.
  • Useful for wrinkle treatment and prevention research.