Learn More
Medchemexpress LLC 7-(4-chlorobenzyl)-1,3-dioxohexahydroimidazo[1,5-a]pyrazin-2(3H)-yl-2,3-dihydro-4H-benzo[b]oxazine-4 | 2331255-53-7 | >98.0% | 50 MG
SDP
Catalog No. 5000231987
Encompass_Preferred
ABC99 is an N-hydroxyhydantoin carbamate that selectively and irreversibly inhibits NOTUM, the Wnt-deacylating serine hydrolase; it preserves Wnt3A signaling in cell-based assays and is used as a chemical probe to study Wnt pathway modulation.
  • Selective NOTUM inhibitor with IC50 13 nM.
  • Preserves Wnt3A signaling in cell-based reporter assays (EC50 89 nM).
  • N-hydroxyhydantoin carbamate chemical class.
  • Molecular weight 456.9 g/mol; formula C22H21ClN4O5.
  • Supplied for research use in in vitro studies.
Catalog No. 50-002-31987 Supplier Medchemexpress LLC Supplier No. HY12283250MG
ABC99 is an N-hydroxyhydantoin carbamate that selectively and irreversibly inhibits NOTUM, the Wnt-deacylating serine hydrolase; it preserves Wnt3A signaling in cell-based assays and is used as a chemical probe to study Wnt pathway modulation.
  • Selective NOTUM inhibitor with IC50 13 nM.
  • Preserves Wnt3A signaling in cell-based reporter assays (EC50 89 nM).
  • N-hydroxyhydantoin carbamate chemical class.
  • Molecular weight 456.9 g/mol; formula C22H21ClN4O5.
  • Supplied for research use in in vitro studies.