Learn More
Cayman Chemical 7 4-DI-O-methylapIgenIn 10mg
SDP
Catalog No. 502909938
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits MAO-B but not MAO-A (IC50s = 0.198 and >100 µM, respectively), as well as p38α MAPK but not JNK3 (IC50s = 27.8 and >100 µM, respectively); inhibits NO production in LPS-stimulated RAW 264.7 macrophages (IC50 = 24.5 µM); reduces the growth of C. albicans at 45 UG/ml; inhibits carrageenan-induced edema in rats (ED25 = 75 mg/kg)
Catalog No. 50-290-9938 Supplier Cayman Chemical Supplier No. 3538810 MG
A flavonoid with diverse biological activities; inhibits MAO-B but not MAO-A (IC50s = 0.198 and >100 µM, respectively), as well as p38α MAPK but not JNK3 (IC50s = 27.8 and >100 µM, respectively); inhibits NO production in LPS-stimulated RAW 264.7 macrophages (IC50 = 24.5 µM); reduces the growth of C. albicans at 45 UG/ml; inhibits carrageenan-induced edema in rats (ED25 = 75 mg/kg)