Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7,4'-Dihydroxyflavone | 2196-14-7 | 25 MG
SDP

Catalog No. 5000421595
Encompass_Preferred

7,4'-Dihydroxyflavone (7,4'-DHF) is a flavonoid isolated from Glycyrrhiza uralensis. It functions as an eotaxin/CCL11 inhibitor and a CBR1 inhibitor with an IC50 of 0.28 μM. This compound suppresses eotaxin production and prevents dexamethasone (Dex)-paradoxical adverse effects. Furthermore, 7,4'-DHF inhibits MUC5AC gene expression, mucus production, and secretion by regulating NF-κB, STAT6, and HDAC2, decreasing PMA-stimulated MUC5AC expression in NCI-H292 human airway epithelial cells with an IC50 of 1.4 μM.

  • Purity: 99.64%
  • Molecular weight: 254.24
  • Formula: C15H10O4
  • CAS number: 2196-14-7
  • Appearance: Solid, light yellow to yellow
  • Structure classification: Flavonoids, Flavones, Phenols, Polyphenols
  • Acts as eotaxin/CCL11 and CBR1 inhibitor (IC50: 0.28 μM)
  • Inhibits MUC5AC gene expression and mucus production
  • IC50: 1.4 μM (NCI-H292 human airway epithelial cell)

Catalog No. 50-004-21595 Supplier Medchemexpress LLC Supplier No. 25MGHYN2609
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7,4'-Dihydroxyflavone (7,4'-DHF) is a flavonoid isolated from Glycyrrhiza uralensis. It functions as an eotaxin/CCL11 inhibitor and a CBR1 inhibitor with an IC50 of 0.28 μM. This compound suppresses eotaxin production and prevents dexamethasone (Dex)-paradoxical adverse effects. Furthermore, 7,4'-DHF inhibits MUC5AC gene expression, mucus production, and secretion by regulating NF-κB, STAT6, and HDAC2, decreasing PMA-stimulated MUC5AC expression in NCI-H292 human airway epithelial cells with an IC50 of 1.4 μM.

  • Purity: 99.64%
  • Molecular weight: 254.24
  • Formula: C15H10O4
  • CAS number: 2196-14-7
  • Appearance: Solid, light yellow to yellow
  • Structure classification: Flavonoids, Flavones, Phenols, Polyphenols
  • Acts as eotaxin/CCL11 and CBR1 inhibitor (IC50: 0.28 μM)
  • Inhibits MUC5AC gene expression and mucus production
  • IC50: 1.4 μM (NCI-H292 human airway epithelial cell)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.