Learn More
Medchemexpress LLC 7,4'-Dihydroxyflavone | 2196-14-7 | MFCD00017694 | 1 ML
SDP
Catalog No. 5000420827
Encompass_Preferred
7,4'-Dihydroxyflavone (7,4'-DHF) is a flavonoid isolated from *Glycyrrhiza uralensis*. It acts as an eotaxin/CCL11 inhibitor and a CBR1 inhibitor with an IC50 of 0.28 μM. This compound can consistently suppress eotaxin production and prevent dexamethasone (Dex)‐paradoxical adverse effects on eotaxin production.
  • Inhibits MUC5AC gene expression
  • Reduces mucus production and secretion
  • Regulates NF-κB, STAT6, and HDAC2 pathways
  • Decreases phorbol 12-myristate 13-acetate (PMA) stimulated NCI-H292 human airway epithelial cell MUC5AC gene expression and mucus production with an IC50 value of 1.4 μM
Catalog No. 50-004-20827 Supplier Medchemexpress LLC Supplier No. HYN260910MM/1ML
7,4'-Dihydroxyflavone (7,4'-DHF) is a flavonoid isolated from *Glycyrrhiza uralensis*. It acts as an eotaxin/CCL11 inhibitor and a CBR1 inhibitor with an IC50 of 0.28 μM. This compound can consistently suppress eotaxin production and prevent dexamethasone (Dex)‐paradoxical adverse effects on eotaxin production.
  • Inhibits MUC5AC gene expression
  • Reduces mucus production and secretion
  • Regulates NF-κB, STAT6, and HDAC2 pathways
  • Decreases phorbol 12-myristate 13-acetate (PMA) stimulated NCI-H292 human airway epithelial cell MUC5AC gene expression and mucus production with an IC50 value of 1.4 μM