Learn More
TARGETMOL CHEMICALS INC 7 4 -Dihydroxyflavone 25MG
SDP
Catalog No. 502877080
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg and bulk. Please contact Fisher for quotes. 7,4'-Dihydroxyflavone (4',7-Dihydroxyflavone) can induce transcription of nodulation (nod) genes in Rhizobium meliloti. It has inhibitory activities against COX-2. Purity 99.6%
Catalog No. 50-287-7080 Supplier TARGETMOL CHEMICALS INC Supplier No. TN127325MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg and bulk. Please contact Fisher for quotes. 7,4'-Dihydroxyflavone (4',7-Dihydroxyflavone) can induce transcription of nodulation (nod) genes in Rhizobium meliloti. It has inhibitory activities against COX-2. Purity 99.6%