Learn More
CHEMSCIENCE Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 7.5, Individually Serialized, Tolerance ± 0.01 mL
Catalog No. NC2318870
Encompass_Preferred
Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 7.5, Individually Serialized, Tolerance ± 0.01 mL
Catalog No. NC2318870 Supplier CHEMSCIENCE Supplier No. 123.302.07A
Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 7.5, Individually Serialized, Tolerance ± 0.01 mL