Learn More
Teknova 7.5M Ammonium Acetate Solution. 250mL, Sterile 4 pack.
SDP
Catalog No. 502387587
Encompass
Ammonium sulfate is a powerful inorganic disinfectant tbat can be used as a precipitating agent for both proteins and plasmids.
Catalog No. 50-238-7587 Supplier Teknova Supplier No. A20004PK
Ammonium sulfate is a powerful inorganic disinfectant tbat can be used as a precipitating agent for both proteins and plasmids.