Learn More
Uline 7 1/2 x 15"" White Block Whirl-Pak Bags - 69oz
Catalog No. NC2241626
Encompass
7 1?2 x 15" White Block Whirl-Pak¿ Bags - 69ozCollect soil and specimen samples in super-strong blended polyethylene.Whirl bag shut, fold ends to create airtight, leakproof seal.Sterile. Meets FDA and USDA specifications.Write-on area for quick, easy labeling.
This item is not returnable. View return policy
Catalog No. NC2241626 Supplier Uline Supplier No. S22730
7 1?2 x 15" White Block Whirl-Pak¿ Bags - 69ozCollect soil and specimen samples in super-strong blended polyethylene.Whirl bag shut, fold ends to create airtight, leakproof seal.Sterile. Meets FDA and USDA specifications.Write-on area for quick, easy labeling.