Learn More
Analytical Sales & Services 7/64” Hex Bit for Power Tools
Catalog No. 5000807234
Encompass
7/64” Hex Bit for Power Tools\n\n• 7/64" Hex Bit\n• 1/4" Hex Shank\n• Compatible with Analytical’s Milwaukee battery powered screwdriver\n• Use with Radiochemistry Reactors (96950) or Conversion Kit (96940)\n\nQty/Pkg: Each
Catalog No. 50-008-07234 Supplier Analytical Sales & Services Supplier No. HB764
7/64” Hex Bit for Power Tools\n\n• 7/64" Hex Bit\n• 1/4" Hex Shank\n• Compatible with Analytical’s Milwaukee battery powered screwdriver\n• Use with Radiochemistry Reactors (96950) or Conversion Kit (96940)\n\nQty/Pkg: Each