Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7,8,3',4'-Tetrahydroxyflavone | 3440-24-2 | 10 MG
SDP

Catalog No. 5000418164
Encompass_Preferred

7,8,3′,4′-Tetrahydroxyflavone (compound 2) is a potent xanthine oxidase (XOD) inhibitor with an IC50 value of 10.488 μM. It is intended for research use only and is not sold to patients. The product is a light yellow to yellow solid with a molecular weight of 286.24 and a formula of C15H10O6. Its structure is classified under Flavonoids and Flavones, and its initial source is identified as Plants Leguminosae Acacia confusa Merr.

  • Potent xanthine oxidase (XOD) inhibitor (IC50: 10.488 μM).
  • High purity (99.51%).
  • Appearance: light yellow to yellow solid.
  • Soluble in DMSO (200 mg/mL).
  • In vivo protocols available for clear solutions.
  • Solid storage: 4°C, protected from light, under nitrogen.
  • Documentation includes data sheet, COA, SDS, and handling instructions.
  • Cited in publications for xanthine oxidase inhibition research.

Catalog No. 50-004-18164 Supplier Medchemexpress LLC Supplier No. HYN860010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7,8,3′,4′-Tetrahydroxyflavone (compound 2) is a potent xanthine oxidase (XOD) inhibitor with an IC50 value of 10.488 μM. It is intended for research use only and is not sold to patients. The product is a light yellow to yellow solid with a molecular weight of 286.24 and a formula of C15H10O6. Its structure is classified under Flavonoids and Flavones, and its initial source is identified as Plants Leguminosae Acacia confusa Merr.

  • Potent xanthine oxidase (XOD) inhibitor (IC50: 10.488 μM).
  • High purity (99.51%).
  • Appearance: light yellow to yellow solid.
  • Soluble in DMSO (200 mg/mL).
  • In vivo protocols available for clear solutions.
  • Solid storage: 4°C, protected from light, under nitrogen.
  • Documentation includes data sheet, COA, SDS, and handling instructions.
  • Cited in publications for xanthine oxidase inhibition research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.