Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7,8-Dihydro-5H-cyclohepta[b]pyridine-5,9(6H)-dione | 39713-40-1 | 99.0% | C10H9NO2 | 100 G
SDP

Catalog No. 5000434417
Encompass_Preferred

7,8-Dihydro-5H-cyclohepta[b]pyridine-5,9(6H)-dione (CAS 39713-40-1) is a chemical compound with the molecular formula C10H9NO2 and a molecular weight of 175.19. This solid appears brown to reddish brown and is typically used as a laboratory chemical. It is stable under recommended storage conditions and has a purity of 99.0% as determined by LCMS. This product is for research use only.

  • Chemical formula: C10H9NO2
  • Molecular weight: 175.19
  • Appearance: brown to reddish brown solid
  • Purity: 99.0% (LCMS)
  • Stable under recommended storage conditions
  • Intended for laboratory and research use

Catalog No. 50-004-34417 Supplier Medchemexpress LLC Supplier No. HY79058100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7,8-Dihydro-5H-cyclohepta[b]pyridine-5,9(6H)-dione (CAS 39713-40-1) is a chemical compound with the molecular formula C10H9NO2 and a molecular weight of 175.19. This solid appears brown to reddish brown and is typically used as a laboratory chemical. It is stable under recommended storage conditions and has a purity of 99.0% as determined by LCMS. This product is for research use only.

  • Chemical formula: C10H9NO2
  • Molecular weight: 175.19
  • Appearance: brown to reddish brown solid
  • Purity: 99.0% (LCMS)
  • Stable under recommended storage conditions
  • Intended for laboratory and research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.