Learn More
Cayman Chemical 7 8-dIhydro-L-BIopterIn 10mg
SDP
Catalog No. 502896268
Encompass_Preferred
BH2 is a precursor of BH4 synthesis. It is a noncompetitive inhibitor of GTP cyclohydrolase I, with a Ki of 14.4 µM.{1268}
Catalog No. 50-289-6268 Supplier Cayman Chemical Supplier No. 8188210 MG
BH2 is a precursor of BH4 synthesis. It is a noncompetitive inhibitor of GTP cyclohydrolase I, with a Ki of 14.4 µM.{1268}