Learn More
Medchemexpress LLC HY-W008646 5mg Medchemexpress, 7,8-Dihydro-L-biopterin CAS:6779-87-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0086465MG
Encompass_Preferred
Medchemexpress, HY-W008646 5mg 7,8-Dihydro-L-biopterin CAS:6779-87-9 7,8-Dihydro-L-biopterin is an oxidation product of tetrahydrobiopterin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4520