Learn More
Medchemexpress LLC 7,8-Dihydro-L-biopterin | 6779-87-9 | 99.79% | 239.23 | 100 MG
SDP
Catalog No. 5000452569
Encompass_Preferred
7,8-Dihydro-L-biopterin is a NOS uncoupling inducer that can cross the blood-brain barrier. It is a reduced non-conjugated pteridine and the main metabolite of 4-amino-tetrahydro-L-biopterin, which undergoes photooxidation to form biopterin. This compound promotes the conversion of nitric oxide synthase to a superoxide-producing form, leading to increased oxidative stress in the renal outer medulla and inducing apoptosis.
  • NOS uncoupling inducer
  • Can cross the blood-brain barrier
  • Promotes conversion of nitric oxide synthase to a superoxide-producing form
  • Increases oxidative stress in the renal outer medulla
  • Induces apoptosis
  • Sensitive to inhibitory effect of SOD
  • Used in research related to salt-sensitive hypertension and neurodegenerative diseases
Catalog No. 50-004-52569 Supplier Medchemexpress LLC Supplier No. HYW008646100MG
7,8-Dihydro-L-biopterin is a NOS uncoupling inducer that can cross the blood-brain barrier. It is a reduced non-conjugated pteridine and the main metabolite of 4-amino-tetrahydro-L-biopterin, which undergoes photooxidation to form biopterin. This compound promotes the conversion of nitric oxide synthase to a superoxide-producing form, leading to increased oxidative stress in the renal outer medulla and inducing apoptosis.
  • NOS uncoupling inducer
  • Can cross the blood-brain barrier
  • Promotes conversion of nitric oxide synthase to a superoxide-producing form
  • Increases oxidative stress in the renal outer medulla
  • Induces apoptosis
  • Sensitive to inhibitory effect of SOD
  • Used in research related to salt-sensitive hypertension and neurodegenerative diseases