Learn More
Selleck Chemical LLC 7 8-Dihydroxyflavone
SDP
Catalog No. 5000000889
Encompass_Preferred
7 8-Dihydroxyflavone (7 8-DHF) acts as a potent and selective small-molecule agonist of the TrkB receptor (Kd 320 nM) the main signaling receptor of brain-derived neurotrophic factor (BDNF)
Catalog No. 50-000-00889 Supplier Selleck Chemical LLC Supplier No. S831910MM/1ML
7 8-Dihydroxyflavone (7 8-DHF) acts as a potent and selective small-molecule agonist of the TrkB receptor (Kd 320 nM) the main signaling receptor of brain-derived neurotrophic factor (BDNF)