Learn More
Apexbio Technology LLC 7,8-Dihydroxyflavone 38183-03-8 10mg
SDP
Catalog No. 5000089080
Encompass_Preferred
7 8-Dihydroxyflavone (CAS 38183-03-8) is a small-molecule compound that acts as a selective agonist of the tropomyosin receptor kinase B (TrkB) By mimicking the action of brain-derived neurotrophic factor (BDNF) it activates TrkB-mediated signaling pathways resulting in enhanced neuronal survival and modulation of synaptic plasticity 7 8-Dihydroxyflavone is widely utilized in experimental studies on neurodegenerative diseases and neurological disorders serving as a valuable tool for investigating neurotrophic signaling mechanisms and the development of potential therapeutics for central nervous system pathologies
Catalog No. 50-000-89080 Supplier Apexbio Technology LLC Supplier No. B548410
7 8-Dihydroxyflavone (CAS 38183-03-8) is a small-molecule compound that acts as a selective agonist of the tropomyosin receptor kinase B (TrkB) By mimicking the action of brain-derived neurotrophic factor (BDNF) it activates TrkB-mediated signaling pathways resulting in enhanced neuronal survival and modulation of synaptic plasticity 7 8-Dihydroxyflavone is widely utilized in experimental studies on neurodegenerative diseases and neurological disorders serving as a valuable tool for investigating neurotrophic signaling mechanisms and the development of potential therapeutics for central nervous system pathologies