Learn More
Medchemexpress LLC 7,8-Dihydroxyflavone | 38183-03-8 | 254.24 | 25 MG
SDP
Catalog No. 5000366827
Encompass_Preferred
7,8-Dihydroxyflavone is a potent and selective TrkB agonist that mimics the physiological actions of Brain-derived neurotrophic factor (BDNF). It displays therapeutic efficacy toward various neurological diseases.
  • Protects primary cortical neurons and locus coeruleus neurons from Aβ-induced toxicity
  • Promotes dendritic growth and synaptogenesis
  • Prevents synaptic loss and memory deficits in an Alzheimer's Disease model
  • Activates TrkB in the brain
  • Inhibits kainic acid-induced toxicity
  • Decreases infarct volumes in stroke
  • Neuroprotective in a Parkinson disease model
Catalog No. 50-003-66827 Supplier Medchemexpress LLC Supplier No. 25MGHYW013372
7,8-Dihydroxyflavone is a potent and selective TrkB agonist that mimics the physiological actions of Brain-derived neurotrophic factor (BDNF). It displays therapeutic efficacy toward various neurological diseases.
  • Protects primary cortical neurons and locus coeruleus neurons from Aβ-induced toxicity
  • Promotes dendritic growth and synaptogenesis
  • Prevents synaptic loss and memory deficits in an Alzheimer's Disease model
  • Activates TrkB in the brain
  • Inhibits kainic acid-induced toxicity
  • Decreases infarct volumes in stroke
  • Neuroprotective in a Parkinson disease model