Learn More
Gelest Inc 7.0 to 8.0% Vinylmethylsiloxane-dimethylsiloxane copolymer | Trimethylsiloxy terminated | 800-1, 200cs | CAS No.: 67762-94-1 | 100g
Supplier:  Gelest Inc VDT731100G
Encompass
This item has a minimum order quantity of 2 per supplier requirements
7.0 to 8.0% Vinylmethylsiloxane-dimethylsiloxane copolymer | Trimethylsiloxy terminated | 800-1, 200cs | CAS No.: 67762-94-1 | 100g
Catalog No. NC9075865