Learn More
Aat Bioquest 7-AAD [7-Aminoactinomycin D] CAS 7240-37-1 (1 mg)
SDP
Supplier:  Aat Bioquest 17501
Encompass_Preferred
7-Amino actinomycin D 7-AAD is a non-permeant dye that can be used to identify non-viable cells. This catalog is for 1 mg.
Catalog No. 50-224-9711