Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Acetoxy-4-bromomethylcoumarin 98.0+%, TCI America™
SDP

Catalog No. A09791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A09791G Supplier TCI America Supplier No. A09791G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4-8-2747
Molecular Formula C12H9BrO4
Molecular Weight (g/mol) 297.10
MDL Number MFCD00010707
InChI Key YDJFMNSSDOXBSR-UHFFFAOYSA-N
PubChem CID 188296
IUPAC Name [4-(bromomethyl)-2-oxochromen-7-yl] acetate
SMILES CC(=O)OC1=CC2=C(C=C1)C(=CC(=O)O2)CBr

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 297.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Acetoxy-4-bromomethylcoumarin [for HPLC Labeling]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.