Learn More
TARGETMOL CHEMICALS INC 7-ACETOXY-4-METHYLCOUMAR 25MG
SDP
Catalog No. 502576362
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 7-Acetoxy-4-methylcoumarin is a fluorogenic substrate for carboxylesterase yielding a blue fluorescent solution upon cleavage. The compound has been used in aprotic media a cell membrane environmental mimic to study superoxide and related oxyl-radical reactions of model biological membranes dispersed in aqueous media. Due to the ability for AchE to cleave and activate 7-Acetoxy-4-methylcoumarin the fluorogenic substrate has been used to study the AchE inhibitor Tacrine·HCl (sc-200172). The compound has also been isolated from Trigonella foenum graceum. purity: 88%
Catalog No. 50-257-6362 Supplier TARGETMOL CHEMICALS INC Supplier No. T703725MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 7-Acetoxy-4-methylcoumarin is a fluorogenic substrate for carboxylesterase yielding a blue fluorescent solution upon cleavage. The compound has been used in aprotic media a cell membrane environmental mimic to study superoxide and related oxyl-radical reactions of model biological membranes dispersed in aqueous media. Due to the ability for AchE to cleave and activate 7-Acetoxy-4-methylcoumarin the fluorogenic substrate has been used to study the AchE inhibitor Tacrine·HCl (sc-200172). The compound has also been isolated from Trigonella foenum graceum. purity: 88%