Learn More
Medchemexpress LLC 7-Acetoxy-4-methylcoumarin | 2747-05-9 | 98.7% | 218.21 | 1 G
SDP
Catalog No. 5000351480
Encompass_Preferred
7-Acetoxy-4-methylcoumarin is an inhibitor of glutathione S-transferase (GST). This compound inhibits AFB1-DNA binding in vitro with 36.7% inhibition.
  • Inhibits glutathione S-transferase.
  • Inhibits AFB1-DNA binding in vitro with 36.7% inhibition.
  • Suitable for research use only.
Catalog No. 50-003-51480 Supplier Medchemexpress LLC Supplier No. HY1378771G
7-Acetoxy-4-methylcoumarin is an inhibitor of glutathione S-transferase (GST). This compound inhibits AFB1-DNA binding in vitro with 36.7% inhibition.
  • Inhibits glutathione S-transferase.
  • Inhibits AFB1-DNA binding in vitro with 36.7% inhibition.
  • Suitable for research use only.