Learn More
Medchemexpress LLC 7-Acetoxy-4-methylcoumarin | 2747-05-9 | 98.7% | 218.21 | 10 G
SDP
Catalog No. 5000350588
Encompass_Preferred
7-Acetoxy-4-methylcoumarin is a chemical compound known for its biological activity as an inhibitor of Glutathione S-transferase (GST). It has demonstrated inhibitory effects on AFB1-DNA binding in vitro.
  • Inhibits Glutathione S-transferase (GST).
  • Inhibits AFB1-DNA binding in vitro with 36.7% inhibition.
Catalog No. 50-003-50588 Supplier Medchemexpress LLC Supplier No. HY13787710G
7-Acetoxy-4-methylcoumarin is a chemical compound known for its biological activity as an inhibitor of Glutathione S-transferase (GST). It has demonstrated inhibitory effects on AFB1-DNA binding in vitro.
  • Inhibits Glutathione S-transferase (GST).
  • Inhibits AFB1-DNA binding in vitro with 36.7% inhibition.