Learn More
Medchemexpress LLC 7-acetoxy-4-methylcoumarin | 2747-05-9 | 98.7% | 218.21 | 10 MM 1 ML
SDP
Catalog No. 5000349647
Encompass_Preferred
7-Acetoxy-4-methylcoumarin, also known as 4-Methylumbelliferyl acetate, is a chemical compound that functions as an inhibitor of Glutathione S-transferase (GST).
  • Acts as an inhibitor of glutathione S-transferase
  • Inhibits AFB1-DNA binding in vitro with a 36.7% inhibition rate
  • Appears as a white to light yellow solid
  • Soluble in DMSO at 25 mg/mL (114.57 mM)
  • For long-term storage, keep powder at -20°C for up to 3 years or 4°C for up to 2 years.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Intended for research use only
Catalog No. 50-003-49647 Supplier Medchemexpress LLC Supplier No. HY13787710MM/1ML
7-Acetoxy-4-methylcoumarin, also known as 4-Methylumbelliferyl acetate, is a chemical compound that functions as an inhibitor of Glutathione S-transferase (GST).
  • Acts as an inhibitor of glutathione S-transferase
  • Inhibits AFB1-DNA binding in vitro with a 36.7% inhibition rate
  • Appears as a white to light yellow solid
  • Soluble in DMSO at 25 mg/mL (114.57 mM)
  • For long-term storage, keep powder at -20°C for up to 3 years or 4°C for up to 2 years.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Intended for research use only