Learn More
Medchemexpress LLC 7-Acetoxy-4-methylcoumarin | 2747-05-9 | 98.7% | 218.21 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY137877100MG
Encompass_Preferred
7-Acetoxy-4-methylcoumarin, also known as 4-Methylumbelliferyl acetate, is an inhibitor of Glutathione S-transferase (GST). It has been shown to inhibit AFB1-DNA binding in vitro with 36.7% inhibition. This product appears as a white to light yellow solid with the chemical formula C12H10O4 and is intended for research use only.
  • Inhibitor of GST
  • Inhibits AFB1-DNA binding in vitro with 36.7% inhibition
  • Solid appearance
  • White to light yellow color
  • For research use only
Catalog No. 50-003-51478