Learn More
Medchemexpress LLC 7-Acetoxy-4-methylcoumarin | 2747-05-9 | 98.7% | 218.21 | 5 G
SDP
Catalog No. 5000347987
Encompass_Preferred
7-Acetoxy-4-methylcoumarin is an inhibitor of Glutathione S-transferase (GST). It is used in various research applications and is provided as a white to light yellow solid.
  • Inhibits AFB1-DNA binding in vitro
  • Shows 36.7% inhibition of AFB1-DNA binding
  • Supplied as a solid
  • Color ranges from white to light yellow
Catalog No. 50-003-47987 Supplier Medchemexpress LLC Supplier No. HY1378775G
7-Acetoxy-4-methylcoumarin is an inhibitor of Glutathione S-transferase (GST). It is used in various research applications and is provided as a white to light yellow solid.
  • Inhibits AFB1-DNA binding in vitro
  • Shows 36.7% inhibition of AFB1-DNA binding
  • Supplied as a solid
  • Color ranges from white to light yellow