Learn More
Medchemexpress LLC 7-Acetoxy-4-methylcoumarin | 2747-05-9 | 98.7% | 218.21 | 50 G
SDP
Catalog No. 5000350681
Encompass_Preferred
7-Acetoxy-4-methylcoumarin is an inhibitor of Glutathione S-transferase (GST) that has been shown to inhibit AFB1-DNA binding in vitro with 36.7% inhibition. This compound is intended for research use only.
  • Inhibits glutathione S-transferase (GST)
  • Inhibits AFB1-DNA binding in vitro
  • For research use only
Catalog No. 50-003-50681 Supplier Medchemexpress LLC Supplier No. HY13787750G
7-Acetoxy-4-methylcoumarin is an inhibitor of Glutathione S-transferase (GST) that has been shown to inhibit AFB1-DNA binding in vitro with 36.7% inhibition. This compound is intended for research use only.
  • Inhibits glutathione S-transferase (GST)
  • Inhibits AFB1-DNA binding in vitro
  • For research use only