Learn More
Cayman Chemical 7-acetyl Paclitaxel, Purity: ≥98%, CAS: 92950-39-5, 1 mg
SDP
Catalog No. 502918586
Encompass_Preferred
An inhibitor of microtubule depolymerization; inhibits calcium-induced microtubule depolymerization at 10 µM; reduces the growth of J774.2 macrophages (IC50 = ~60 nM)
Catalog No. 50-291-8586 Supplier Cayman Chemical Supplier No. 392881 MG
An inhibitor of microtubule depolymerization; inhibits calcium-induced microtubule depolymerization at 10 µM; reduces the growth of J774.2 macrophages (IC50 = ~60 nM)