Learn More
MOLECULAR DEPOT LLC 7-Alkynyl-Fucose - 0.1 mg
Catalog No. 5000013354
Encompass_Preferred
7-Alkynyl-Fucose is a derivative of fucose used as a high sensitivity research imaging probe in research as well as other applications in cellular biology It has a molecular weight of 356 32 g/mol and is supplied as an amorphous white powder This product is for in vitro research use only This product is not for human or animal use
Catalog No. 50-000-13354 Supplier MOLECULAR DEPOT LLC Supplier No. MDP0885
Add to Cart
Add to Cart
7-Alkynyl-Fucose is a derivative of fucose used as a high sensitivity research imaging probe in research as well as other applications in cellular biology It has a molecular weight of 356 32 g/mol and is supplied as an amorphous white powder This product is for in vitro research use only This product is not for human or animal use