Learn More
Accela Chembio Inc 7-amino-1h-indazole | 5g | 21443-96-9 | MFCD00022790 | 97+% | Shelf Life: 1260 Days | Light Sensitive/n2/+4
Catalog No. 50250301
Encompass_Preferred
7-amino-1h-indazole | 5g | 21443-96-9 | MFCD00022790 | 97+% | Shelf Life: 1260 Days | Light Sensitive/n2/+4
Catalog No. 50-250-301 Supplier Accela Chembio Inc Supplier No. SY0038965G
7-amino-1h-indazole | 5g | 21443-96-9 | MFCD00022790 | 97+% | Shelf Life: 1260 Days | Light Sensitive/n2/+4