Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-amino-4-methyl coumarin-arachidonamide | 862913-13-1 | 99.5% | 5 MG
SDP

Catalog No. 5000234023
Encompass_Preferred

AMC Arachidonoyl Amide is a fluorogenic substrate for measuring fatty acid amide hydrolase (FAAH) activity. It links an arachidonoyl fatty acyl chain to a 7-amino-4-methylcoumarin (AMC) fluorophore and is supplied as a high-purity research reagent.

  • Used to measure FAAH activity by release of a fluorescent AMC reporter.
  • Contains an AMC fluorophore linked to an arachidonoyl chain for substrate specificity.
  • High purity suitable for biochemical assays (HPLC 99.5%).
  • Recommended storage at -80°C to maintain stability.
  • Molecular weight 461.64 g/mol; formula C30H39NO3.

Catalog No. 50-002-34023 Supplier Medchemexpress LLC Supplier No. HY1209575MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AMC Arachidonoyl Amide is a fluorogenic substrate for measuring fatty acid amide hydrolase (FAAH) activity. It links an arachidonoyl fatty acyl chain to a 7-amino-4-methylcoumarin (AMC) fluorophore and is supplied as a high-purity research reagent.

  • Used to measure FAAH activity by release of a fluorescent AMC reporter.
  • Contains an AMC fluorophore linked to an arachidonoyl chain for substrate specificity.
  • High purity suitable for biochemical assays (HPLC 99.5%).
  • Recommended storage at -80°C to maintain stability.
  • Molecular weight 461.64 g/mol; formula C30H39NO3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.