Learn More
AdipoGen 7-Amino-4-methylcoumarin
SDP
Supplier:  AdipoGen CDXA0021M250
Encompass_Preferred
Chemical. CAS 26093-31-2. Formula C10H9NO2. MW 175.18. Synthetic. Widely used fluorophore well-known for preparing fluorogenic 7-Amido-4-methylcoumarin AMC substrates for cystine aminopeptidase and other hydrolases. Used as reference compound in enzyme assays.
Catalog No. 50-256-1863