Learn More
Cayman Chemical 7-Amino-4-methylcoumarin, Purity: ≥98%, CAS: 26093-31-2, 250 mg
SDP
Catalog No. 502940000
Encompass_Preferred
A fluorescent probe for peptide labeling commonly used in the study of proteases; conjugation via its amino group to a peptide substrate results in quenching of the fluorescent signal; upon enzymatic cleavage of the peptide by proteases, AMC is released and its fluorescence can be used to quantify enzyme activity; ex/em = 345/445 nm, respectively
Catalog No. 50-294-0000 Supplier Cayman Chemical Supplier No. 27792250 MG
A fluorescent probe for peptide labeling commonly used in the study of proteases; conjugation via its amino group to a peptide substrate results in quenching of the fluorescent signal; upon enzymatic cleavage of the peptide by proteases, AMC is released and its fluorescence can be used to quantify enzyme activity; ex/em = 345/445 nm, respectively