Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Amino-4-methylcoumarin-3-acetic acid | 106562-32-7 | 99.9% | 50 MG
SDP

Supplier:  Medchemexpress LLC HYD002550MG

Encompass_Preferred

7-Amino-4-methylcoumarin-3-acetic acid is a fluorescent protein labelling agent. It emits in the blue region (440-460 nm) on activation with UV light (350 nm).

  • Molecular weight: 233.22
  • Molecular formula: C12H11NO4
  • Appearance: Solid
  • Color: Off-white to light brown
  • Emission (Em): 454
  • Excitation (Ex): 344
  • Target: Fluorescent dye
  • Pathway: Others

Catalog No. 50-004-10609


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.